Learn More
Vimentin Antibody (2D1), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-92687UV
This item is not returnable.
View return policy
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
| Vimentin | |
| Monoclonal | |
| DyLight 350 | |
| P08670 | |
| VIM | |
| Full length recombinant human Vimentin expressed in and purified from E. coli. [UniProt# P08670] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Rat and Mouse and likely all other Mammals. | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
| 2D1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated | |
| FLJ36605, vimentin | |
| Mouse | |
| 50 kDa | |
| 0.1 mL | |
| Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers, Stem Cells | |
| 7431 | |
| Human, Rat, Mouse (Negative) | |
| Purified |