missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Antibody (CL0157) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52867AF350
This item is not returnable.
View return policy
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Gene Knock-Out
Specifications
| Vimentin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin | |
| This antibody was developed using a recombinant protein derived from P08670, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
| 7431 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Gene Knock-Out | |
| CL0157 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction