missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Antibody (RV203), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-97671CL1
This item is not returnable.
View return policy
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Canine,Goat,Hamster,Human,Mouse,Porcine,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| Vimentin | |
| Monoclonal | |
| CoraFluor 1 | |
| epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin | |
| This antibody is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 Mouse myeloma cells with spleen cells from a BALB/c Mouse immunized with a cytoskeletal vimentin extract of calf lens. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
| 7431 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| RV203 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Canine, Goat, Hamster, Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction