missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Antibody (V9), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97670PE
This item is not returnable.
View return policy
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human, Rat, Canine, Chicken, Equine, Feline, Sheep samples. It is validated for Flow Cytometry.
Specifications
| Vimentin | |
| Monoclonal | |
| PE | |
| FLJ36605, vimentin | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| V9 reacts exclusively with vimentin, which is expressed in mesenchymal cells and mesenchymal derived tumors e.g. lymphoma, sarcoma and melanoma. | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| V9 | |
| Flow Cytometry | |
| VIM | |
| Clone V9 is a mouse monoclonal IgG1 antibody derived by fusion of PAI mouse myeloma cells with spleen cells from a BALB/c mouse immunized with vimentin isolated from porcine lens. | |
| 0.1 mL | |
| Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers, Stem Cells | |
| 7431 | |
| Human, Rat, Canine, Chicken, Equine, Equine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction