missing translation for 'onlineSavingsMsg'
Learn More

Vincamine, TRC

Product Code. 30006146
Change view
Click to view available options
Quantity:
2.5 g
10 g
25 g
Unit Size:
10g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006146 25 g 25g
30009167 10 g 10g
17932575 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006146 Supplier Toronto Research Chemicals Supplier No. TRCV31403025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1617-90-9
Molecular Formula C21 H26 N2 O3
Molecular Weight (g/mol) 354.44
Synonym Vincamine, (3α,14β,16α)-14,15-Dihydro-14-hydroxyeburnamenine-14-carboxylic acid methyl ester, Minorine, Eburnamenine-14-carboxylic acid, 14,15-dihydro-14-hydroxy-, methyl ester, (3α,14β,16α)-, Minorine (7CI), Vincamine (6CI,8CI), 1H-Indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine, eburnamenine-14-carboxylic acid deriv., (+)-Vincamine, (+)-cis-Vincamine, Angiopac, Arteriovinca, Devincan, Devinkan, Equipur, Methyl vincaminate, Minorin, NSC 91998, Novicet, Ocu-vinc, Oxybral, Oxygeron, Perval, Pervincamine, Pervone, Sostenil, Tripervan, Vincadar, Vincafarm, Vincafolina, Vincafor, Vincagil, Vincalen, Vincamidol, Vincamin, Vincapan, Vincapront, Vincimax, Vinodrel Retard, Vinvasaunier, Vraap, cis-Vincamine, (12S,13aS,13bS)-13a-Ethyl-2,3,5,6,12,13,13a,13b-octahydro-12-hydroxy-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylic acid methyl ester
IUPAC Name (3α,14β,16α)-14,15-Dihydro-14-hydroxyeburnamenine-14-carboxylic acid methyl ester
SMILES CC[C@@]12CCCN3CCc4c([C@H]13)n(c5ccccc45)[C@](O)(C2)C(=O)OC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C21H26N2O3/c1-3-20-10-6-11-22-12-9-15-14-7-4-5-8-16(14)23(17(15)18(20)22)21(25,13-20)19(24)26-2/h4-5,7-8,18,25H,3,6,9-13H2,1-2H3/t18-,20+,21+/m1/s1
Formula Weight 354.1943
Chemical Name or Material Vincamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.