Learn More
Vinyl Acetate 2000 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30323996
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323996 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30323996 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010383ME
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Vinylacetate
CAS 108-05-4
Analyte or Component Names Vinylacetate
Shipping Condition Ice pack (-20°C)
Molecular Formula C4 H6 O2
Concentration 2000 μg/mL
Certifications/Compliance ISO 17034
Synonym Vinyl Acetate, Acetic acid vinyl ester (8CI), Ethenyl acetate, 1-Acetoxyethylene, Acetic acid, ethenyl ester, Acetoxyethene, Acetoxyethylene, NSC 8404, SN 12T, Vinyl A monomer, Vinyl acetate
InChI Formula InChI=1S/C4H6O2/c1-3-6-4(2)5/h3H,1H2,2H3
SMILES CC(=O)OC=C
IUPAC Name ethenyl acetate
Solvent or Matrix Methanol
Molecular Weight (g/mol) 86.09
Quantity 1 mL
Formula Weight 86.0368 g/mol