Learn More
Brady™ Vinyl with Polyester Overlaminate Durable Marking Tape
Click to view available options
Color:
Black, White
Black, Yellow
Magenta, Yellow
Red, White
Dimensions (L x W):
16.5 m x 25 mm
16.5 m x 38 mm
16.5 m x 50 mm
16.5 m x 75 mm
Unit Size:
1 roll
Specifications
Specifications
| Dimensions (L x W) | 16.5 m x 25 mm |
| Finish | High Gloss |
| For Use With (Application) | Safety marking of obstructions, low hanging objects, pillars, passageways, aisles, and any other potentially hazardous areas or objects |
| Length (Metric) | 16.5 m |
| Packaging Quantity | 1 Roll |
| Temperature Range (Metric) | -40°C to +80°C |
| Thickness | 0.037 mm |
| Width (Metric) | 25 mm |
| Adhesive Type | Rubber-based Pressure-Sensitive |
| Color | Black, Yellow |
| Show More |