missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIP Antibody (2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05163CL1
This item is not returnable.
View return policy
Description
VIP Monoclonal antibody specifically detects VIP in Bovine,Human,Porcine,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| VIP | |
| Monoclonal | |
| CoraFluor 1 | |
| MGC13587, PHM27, vasoactive intestinal peptide, VIP peptides | |
| Vasoactive intestinal peptide | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction | |
| 7432 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| 2 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bovine, Human, Pig, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction