missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIP Monoclonal Antibody (OTI1C3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806855
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the glucagon family. It stimulates myocardial contractility, causes vasodilation, increases glycogenolysis, lowers arterial blood pressure and relaxes the smooth muscle of trachea, stomach and gall bladder. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.Specifications
| VIP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P01282 | |
| VIP | |
| Full length human recombit protein of human VIP produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C3 | |
| Unconjugated | |
| VIP | |
| PHM27 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7432 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction