missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VIP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA126989
This item is not returnable.
View return policy
Description
PA1-26989 detects VIP from human samples.
VIP encodes a protein that belongs to the glucagon family. It stimulates myocardial contractility, causes vasodilation, increases glycogenolysis, lowers arterial blood pressure and relaxes the smooth muscle of trachea, stomach and gall bladder. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications
| VIP | |
| Polyclonal | |
| Unconjugated | |
| VIP | |
| Intestinal peptide PHI-27; Intestinal peptide PHI-42; Intestinal peptide PHM-27; Intestinal peptide PHV-42; MGC13587; PACAP-R-2; Peptide histidine isoleucinamide 27; Peptide histidine methioninamide 27; Peptide histidine valine 42; PHI; PHI, peptide histidine isoleucine; PHM27; prepro-VIP; vasoactive intestinal peptide; Vasoactive intestinal polypeptide; VIP; VIP peptides; vip/phi27; VIP-R-1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7432 | |
| 4°C, do not freeze | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.3 | |
| P01282 | |
| VIP | |
| Synthetic peptide. Exact sequence is proprietary. | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction