missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VIR-2482 Humanized Recombinant Human Monoclonal Antibody (VIR-2482)
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555027
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
VIR-2482 is a monoclonal antibody that targets the hemagglutinin protein of the influenza virus, preventing the virus from binding to and entering host cells.
Specifications
| VIR-2482 Humanized | |
| Recombinant Monoclonal | |
| 3.28 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A/G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA | |
| VIR-2482 | |
| Unconjugated | |
| HA | |
| Recombinant Influenza A virus HA/Hemagglutinin. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction