missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral CMV UL146/vCXC1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF620
This item is not returnable.
View return policy
Description
UL146/vCXC1 Polyclonal specifically detects UL146/vCXC1 in Viral samples. It is validated for Western Blot, Neutralization.
Specifications
| UL146/vCXC1 | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3077566 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 10-30 ug/mL | |
| E. coli-derived recombinant viral CMV UL146/vCXC1 | |
| 100 μg | |
| Primary | |
| Detects viral CMV UL146/vCXC1 in direct ELISAs and Western blots. | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction