missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Viral HCMV UL7 (ABV71537) VersaClone cDNA
Description
- UL7 [Human betaherpesvirus 5]
- UL7 is secreted from HCMV infected human cells and activates Flt-3R with an activity comparable to Flt3L.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | vUL7 |
| Accession Number | ABV71537 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 682 bp |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Virus |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?