missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human PDGF-AA, Animal-Free Protein

Product Code. 18725406
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18725406

Brand: R&D Systems™ AFL221050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. ED50 for this effect is 50 to 200 ng/mL in a fluorometric assay using the redox sensitive dye, Resazurin.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04085.1
For Use With (Application) Bioactivity
Gene ID (Entrez) 5154
Molecular Weight (g/mol) M.W. theroretical: 14 kDa (monomer)
Quantity 50 μg
Source E. coli-derived human PDGF-AA protein Ser87-Thr211, with and without an N-terminal Met Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, ≤ -20°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Gene Alias PDGF1, PDGF-A, PDGFAA
Gene Symbol PDGFA
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 0.2 mg/mL in sterile 4 mM HCl.
Buffer Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Endotoxin Level <0.01 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.