missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Virilizer Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-21363H
This item is not returnable.
View return policy
Description
Virilizer Polyclonal antibody specifically detects Virilizer in Human samples. It is validated for Western Blot
Specifications
| Virilizer | |
| Polyclonal | |
| Western Blot | |
| DKFZP434I116, DKFZp781B2117, fSAP121, functional spliceosome-associated protein 121, KIAA1429, MGC138493, MGC141940, protein virilizer homolog | |
| The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human Virilizer (KIAA1429) using the numbering given in entry NP_056311.2 (GeneID 25962). | |
| 0.1 mL | |
| Cell Biology | |
| 25962 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu