Learn More
Sartorius Stedim Biotech™ Viscose/Polyester Non-Woven Filters, Discs, Grade 2602
Generally used for liquid filtration in environments such as the food processing industry.
Brand:  Sartorius Stedim Biotech™ FT-3-01324-032
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 15932840
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Non-woven
  • Suitable for use in the food processing industry.
  • Application example: Petrochemical industry: Detection of contaminants in dairy products, baby food.
  • Non-sterile, bulk packed.
Specifications
Specifications
150 g/m2
Rayon (Viscose) - Polyester
1000/Pk.
Non-woven Filter
Disc
2602
0.8 mm
32 mm