missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Viscose/Polyester Non-Woven Filters, Discs, Grade 2602
Generally used for liquid filtration in environments such as the food processing industry.
Brand: Sartorius Stedim Biotech™ FT-3-01324-047
Additional Details : Weight : 2.00000kg
Description
- Non-woven
- Suitable for use in the food processing industry.
- Application example: Petrochemical industry: Detection of contaminants in dairy products, baby food.
- Non-sterile, bulk packed.
Specifications
150 g/m2 | |
Rayon (Viscose) - Polyester | |
1000/Pk. | |
Non-woven Filter |
Disc | |
2602 | |
0.8 mm | |
47 mm |