Description
Thermo Scientific™ VISIONlite™ 5 Software enhances the capabilities of our stand-alone Visible and UV-Visible spectrophotometers. The software enables you use your spectrophotometer to its fullest potential, displaying and reporting experimental and analytical results with ease.
Thermo Scientific™ VISIONlite™ ColorCalc software is a powerful stand-alone software package that adds the ability to perform numerous color calculations for a variety of applications. The software records the required transmission or reflectance spectrum and automatically performs calculations using the recorded data.
Thermo Scientific™ VISIONlite™ EnzLab software offers predefined methods and operation procedures for all standard enzymatic test kits including Citric Acid, Lactic Acid, D-Glucose, D-Fructose, Ethanol, Lactose and many more. EnzLab automates and facilitates enzymatic determinations, ensures correct calculations, and provides appropriate analytical reports.
Thermo Scientific™ VISIONlite™ Wine Analysis software allows users to perform an extensive list of predefined photometric, colorimetric and enzymatic analytical procedures common in wine and juice analysis. This PC software features a simple user interface, reliable data recording, and error-free evaluation of results within comprehensive reports.
VISIONlite 5 Software increases the capability of your instrument:
- Add method and data storage for SPECTRONIC 200 Spectrophotometer and GENESYS 20 Visible Spectrophotometer.
- Report results to 0.5nm wavelength resolution and 0.001A for SPECTRONIC 200 Spectrophotometer.
- Perform automated kinetics measurements on SPECTRONIC 200 Spectrophotometer and GENESYS 20 Visible Spectrophotometer.
- Scan on GENESYS 20 Visible Spectrophotometer.
- Produce formatted reports in hard copy or pdf format.
VISIONlite ColorCalc Software turns your GENESYS 10S UV-Vis Spectrophotometer or Thermo Scientific™ Evolution™ 300 UV-Vis Spectrophotometer into a powerful color analyser:
- The Basic package offers all the most common colorspaces, illuminants and observer angles.
- The Advanced package offers specialized calculations for yellowness, pharmaceuticals and quality control in wine production.
Specifications
Specifications
| Type | 6-user Software License |
| For Use With (Equipment) | VISIONlite 5 Software |
| Includes | Includes two copies of the installation disc and a license granting approval for six installed instances of the software |