missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Vismodegib

Description
Vismodegib is a potent hedgehog pathway inhibitor (IC50 = 3 nM). It causes tumor regressions in allograft medulloblastoma mice model and tumor growth inhibition in a xenograft mouse models of colorectal cancer. Vismodegib is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-N-[4-chloro-3-(2-pyridinyl)phenyl]-4-(methylsulfonyl)benzamide |
| CAS | 879085-55-9 |
| Quantity | 10 mg |
| Target | Hedgehog Signaling Inhibitors |
| Molecular Formula | C19H14Cl2N2O3S |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?