missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VISTA/B7-H5/PD-1H Antibody (730801), Alexa Fluor™ 750, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB71262AF750
This item is not returnable.
View return policy
Description
VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Western Blot.
Specifications
| VISTA/B7-H5/PD-1H | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| VSIR | |
| Partial recombinant human VISTA/B7-H5 (amino acids 33-194) [UniProt Q9H7M9] | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot | |
| 730801 | |
| Western Blot | |
| B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
| Mouse | |
| 0.1 mL | |
| 64115 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction