missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VISTA/B7-H5/PD-1H Antibody (855328), Alexa Fluor™ 700, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB9057AF700
This item is not returnable.
View return policy
Description
VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface).
Specifications
| VISTA/B7-H5/PD-1H | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| C10ORF54 | |
| Partial recombinant human VISTA/B7-H5 protein (aa 33-194) | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 855328 | |
| Flow Cytometry, Flow (Cell Surface) | |
| B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, GI24, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 64115 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction