Learn More
Invitrogen™ VISTA Monoclonal Antibody (MIH64), Brilliant Ultra Violet™ 737, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
€ 198.00 - € 360.00
Specifications
| Antigen | VISTA |
|---|---|
| Clone | MIH64 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17423382
|
Invitrogen™
367-1083-80 |
25 μg |
€ 198.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17433382
![]() |
Invitrogen™
367-1083-82 |
100 μg |
€ 360.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This MIH64 monoclonal antibody recognizes mouse VISTA also known as B7-H5 or PD-H1. This MIH64 antibody will work in flow cytometry on both native and paraformaldehyde-fixed cells. Applications Reported: This MIH64 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MIH64 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Brilliant Ultraviolet 737 is a tandem dye that emits at 732 nm and is intended for use on cytometers equipped with an ultraviolet (355 nm) laser.
VISTA Monoclonal antibody specifically detects VISTA in Mouse samples. It is validated for Flow Cytometry.Specifications
| VISTA | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation | |
| VSIR | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| VSIR |
| MIH64 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 737 | |
| Rat | |
| Mouse | |
| Q9D659 | |
| 74048 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.