Learn More
VistaLab™ Technologies Pipette Tips: Non-Sterile
For use with Ovation and MLA Brand pipetters
Brand:  VistaLab™ Technologies 4058-3000
Additional Details : Weight : 0.86180kg
 View more versions of this product
Product Code. 16435104
  • € 635.00 / Pack of 10000
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Description
Description
  • Non-Sterile tips packaged in VistaRak, VistaStak, Stacked Rack, Bulk
  • VistaRak: 5 racks, 192 tips/rack, VistaStak: 3 or 5 Layers of 192 tips, Stacked Rack: 3 or 5 layers of 200 tips, Bulk: 750-1000 tips in a plastic bag or containers
  • Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
  • All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time
Specifications
Specifications
1250 μL
Non-sterile
Blue
3 in.
Macro, Straight
1000 Tips/Bag
No
Non-Filtered
Clear
3.4 in.
Bulk
VistaBulk: Plastic Bag
MLA, Ovation F 200 to 1000 µL, MLA 200 to 1000μL, Ovation