Learn More
VistaLab™ Technologies Pipette Tips: Non-Sterile
For use with Ovation and MLA Brand pipetters
Brand:  VistaLab™ Technologies 9025
Additional Details : Weight : 0.86180kg
 View more versions of this product
Product Code. 16485124
  • € 710.00 / Pack of 10000
Estimated Shipment: 25-07-2024
to see stock.
Add to basket
Description
Description
  • Non-Sterile tips packaged in VistaRak, VistaStak, Stacked Rack, Bulk
  • VistaRak: 5 racks, 192 tips/rack, VistaStak: 3 or 5 Layers of 192 tips, Stacked Rack: 3 or 5 layers of 200 tips, Bulk: 750-1000 tips in a plastic bag or containers
  • Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
  • All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time
Specifications
Specifications
250 μL
Non-sterile
Blue
2.2 in.
Macro, Straight
200 Tips/Layer
MLA, Ovation
Polypropylene
Non-Filtered
Clear
Not Autoclavable
6 in.
Stacked Rack: 5 layers
Single-use