Learn More
Vitamin B12 e-Monocarboxylic Acid, Vitamin B12 e-Monocarboxylic Acid, TRC
Product Code. 30008030
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
10 mg
Unit Size:
0.25mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008030 10 mg 10mg
30005299 1 mg 1mg
17919654 0.25 mg 0.25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30008030 Supplier Toronto Research Chemicals Supplier No. TRCV67565510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Molecular Formula C63H87CoN13O15P
Molecular Weight (g/mol) 1356.35
Synonym Co-(cyano-κC)-cobinic acid-abcdg-pentamide dihydrogen phosphate (ester), inner salt, 3'-ester with (5,6-dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole-κN3), Cobinic acid-abcdg-pentamide, cyanide hydroxide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosylbenzimidazole, Cobinic acid-abcdg-pentamide, cyanide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole, Cobalamin-monocarboxylic acid
SMILES Cl.NC(=N)Nc1ccc(C(=O)Cl)cc1
Specifications
CAS 26264-28-8
Quantity 10 mg
InChI Formula InChI=1S/C62H89N12O15P.CN.Co/c1-29-20-39-40(21-30(29)2)74(28-69-39)57-52(84)53(41(27-75)87-57)89-90(85,86)88-31(3)26-68-48(81)18-19-59(8)37(22-45(65)78)56-62(11)61(10,25-47(67)80)36(13-16-44(64)77)51(73-62)33(5)55-60(9,24-46(66)79)34(12-15-43(63)76)38(70-55)23-42-58(6,7)35(14-17-49(82)83)50(71-42)32(4)54(59)72-56;1-2;/h20-21,23,28,31,34-37,41,52-53,56-57,75,84H,12-19,22,24-27H2,1-11H3,(H14,63,64,65,66,67,68,70,71,72,73,76,77,78,79,80,81,82,83,85,86);;/q;;+2/p-2/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57+,59-,60+,61+,62+;;/m1../s1
Formula Weight 1355.5514
Chemical Name or Material Vitamin B12 e-Monocarboxylic Acid