Learn More
Vitamin D3 Antibody (685503) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB6566I
Product Code. 18574616
  • ONLINE EXCLUSIVE PRICE
    € 664.00
    € 627.90 / 0.10mL
    Save € 36.10
    5% Off
Description
Vitamin D3 Monoclonal antibody specifically detects Vitamin D3 in Broad Species Reactivity samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vitamin D3
Monoclonal
Janelia Fluor 549
50mM Sodium Borate
25-Hydroxyvitamin D3
0.1 mL
Primary
Store at 4°C in the dark.
IgG1
Flow Cytometry (Intracellular Staining), CyTOF
685503
Intracellular Staining by Flow Cytometry, CyTOF-ready
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Broad Species Reactivity