missing translation for 'onlineSavingsMsg'
Learn More

Vitamin D3, MedChemExpress

Product Code. 16406756
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16406756 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16406756 Supplier Medchem Express Supplier No. HY15398
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Vitamin D3 (Cholecalciferol; Colecalciferol) is a naturally occuring form of vitamin D. Vitamin D3 induces cell differentiation and prevents proliferation of cancer cells.

Vitamin D3 (Cholecalciferol

Specifications

Chemical Name or Material Vitamin D3
CAS 67-97-0
Color White
Purity Grade Notes Research
Molecular Formula C27H44O
Quantity 100 mg
Synonym Cholecalciferol Colecalciferol
Solubility Information DMSO : 100 mg/mL (259.98 mM; Need ultrasonic)
SMILES C[C@@]12[C@](CC[C@]2([H])[C@H](C)CCCC(C)C)([H])/C(CCC1)=C/C=C3C(CC[C@H](O)C\3)=C
Molecular Weight (g/mol) 384.64
Formula Weight 384.64
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.