Learn More
Vitamin D4, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 22,23-Dihydroergocalciferol |
| CAS | 511-28-4 |
| Molecular Formula | C28 H46 O |
| Quantity | 10 mg |
| Synonym | Cyclohexanol','4-methylene-3-[(2E)-2-[(1R,3aS,7aR)-octahydro-7a-methyl-1-[(1R,4S)-1,4,5-trimethylhexyl]-4H-inden-4-ylidene]ethylidene]-','(1S,3Z)-, 9,10-Secoergosta-5,7,10(19)-trien-3-ol','(3β,5Z,7E)- (9CI), Ergocalciferol','22,23-dihydro- (6CI), Vitamin D4 (7CI,8CI), (1S,3Z)-4-Methylene-3-[(2E)-2-[(1R,3aS,7aR)-octahydro-7a-methyl-1-[(1R,4S)-1,4,5-trimethylhexyl]-4H-inden-4-ylidene]ethylidene]cyclohexanol, 22,23-Dihydroercalciol, 22,23-Dihydroergocalciferol, 22,23-Dihydrovitamin D2, 24S-Methylcalciol |
| InChI Formula | InChI=1S/C28H46O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h12-13,19-20,22,25-27,29H,4,7-11,14-18H2,1-3,5-6H3/b23-12+,24-13-/t20-,22+,25-,26+,27-,28+/m0/s1 |
| SMILES | CC(C)[C@@H](C)CC[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)CCC3=C)\CCC[C@]12C |
| IUPAC Name | (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(2R,5S)-5,6-dimethylheptan-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol |
| Molecular Weight (g/mol) | 398.66 |
| Formula Weight | 398.3549 |