Learn More
Vitexin-2-O-Rhamnoside, TRC
Specifications
Specifications
| Chemical Name or Material | Vitexin-2''-O-Rhamnoside |
| CAS | 64820-99-1 |
| Molecular Formula | C27 H30 O14 |
| Quantity | 25 mg |
| Synonym | 4H-1-Benzopyran-4-one','8-[2-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]-5,7-dihydroxy-2-(4-hydroxyphenyl)-, 2''-O-Rhamnosylvitexin, 2''-O-α-L-Rhamnopyranosylvitexin, 2''-Rhamnosylvitexin, 8-[2-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Apigenin 8-C-neohesperidoside, Apigenin 8-C-α-L-rhamnopyranosyl-(1→2)-β-D-glucopyranoside], Vitexin 2''-O-rhamnoside, Vitexin 2''-O-α-L-rhamnopyranoside, Vitexin 2''-rhamnoside, Vitexin 2''-α-L-rhamnopyranoside |
| InChI Formula | InChI=1S/C27H30O14/c1-9-19(33)21(35)23(37)27(38-9)41-26-22(36)20(34)16(8-28)40-25(26)18-13(31)6-12(30)17-14(32)7-15(39-24(17)18)10-2-4-11(29)5-3-10/h2-7,9,16,19-23,25-31,33-37H,8H2,1H3/t9-,16+,19-,20+,21+,22-,23+,25-,26+,27-/m0/s1 |
| SMILES | C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2c3c(O)cc(O)c4C(=O)C=C(Oc34)c5ccc(O)cc5)[C@H](O)[C@H](O)[C@H]1O |
| IUPAC Name | 8-[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]-5,7-dihydroxy-2-(4-hydroxyphenyl)chromen-4-one |
| Molecular Weight (g/mol) | 578.52 |
| Formula Weight | 578.1636 |