Learn More
Emma Safety Footwear Vito (Frontier 114) Safety Shoes
Product Code. 15948191
Click to view available options
Size:
39
40
41
42
43
45
46
47
48
Unit Size:
1 pair
Product Code. 15948191
Brand: Emma Safety Footwear MM11409046
Only null left
Add to basket
Add to basket
The executive safety shoe that's made with the same care and  attention to detail associated with  high end luxury shoes. Brazilian 2-tone full grain leather combined with nubuck adds to the stylish look.
  • S3-Safety shoe with sole plate for wet work
  • Thermoplastic Polyurethane (TPU) Outsole
  • Full grain leather construction
  • ESD certified
  • Chrome-free lining
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Black
Design Vito (Frontier 114)
Liner Material Hydro-Tec™ Sanitized Silver & Wellness leather
Material Leather, Nubuck
Size 46
Toe Material Steel