missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Viton™ L/S™ 2-Stop Pump Tubing

Good for food and lab applications

Brand:  Masterflex™ 96428-15

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11789659

  • € 233.00 / Pack of 4
Estimated Shipment: 04-07-2024
to see stock.
Add to basket

Description

Description

  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Food or laboratory applications where FDA compliance is required, acid and solvent transfer, high-temperature work
  • Good chemical resistance, opaque black
  • For use with Masterflex™ multichannel pump heads
Specifications

Specifications

Stopped Pump Tubing
4.8 mm
1.25 ft.
0°C
15
3/16 in.
150°C
FDA-Compliant
Viton
0.38 m
1 / Pk.
1000 mL/min
660 mm Hg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers