missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Viton O Rings

Product Code. 15819512 Shop All Bürkle Products
Change view
Click to view available options
Material:
EPDM
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
15819512 EPDM Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819512 Supplier Buerkle™ Supplier No. 00065044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with Buerkle™ Thread adapters

  • Viton O Rings for several Buerkle™ Thread adapters
  • Spare part
TRUSTED_SUSTAINABILITY

Specifications

Product Type O-Ring
Material EPDM
Color Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.