Learn More
Ismatec™ Viton™ Tubing for Peristaltic Pumps
Click to view available options
Tubing Size:
26
30
42
46
48
Diameter (Metric) Inner:
0.89 mm
1.14 mm
2.06 mm
2.54 mm
2.79 mm
Unit Size:
50ft
Each
Description
- Tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Regulatory approvals to match your unique application
Specifications
Specifications
| Certifications/Compliance | FDA approved; Prop 65 NA (no Prop 65 chemicals) |
| Diameter (Metric) Inner | 2.54 mm |
| Material | Viton |
| Overall Length (English) | 50 ft. |
| Overall Length (Metric) | 15.2 m |
| For Use With | C/L Pump Series |
| Min. Operating Temperature (Metric) | 0°C |
| Tubing Size | 46 |
| Hose Barb | 1/8 in. |
| Maximum Pressure | 25.3 psi, 1.75 bar |
| Show More |