missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ VitroEase™ Methylamine Vanadate Negative Stain

Product Code. 17460703
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17460703 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17460703 Supplier Thermo Scientific™ Supplier No. A51037

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An easy, ready-to-use negative stain for electron microscopy (EM) analysis.

  • An easy, ready-to-use negative stain for electron microscopy (EM) analysis
  • Provided as a stable, ready-to-use 2% stain solution
  • Easy to use with straight-forward staining protocol
  • Excellent spreading properties at a near-neutral pH of 8
  • Uniform, electron-dense stain for EM
  • Produces a light, uniform negative stain for high-resolution fine structure visualization 
  • Dries without crystallization, ideal for EM grids
  • Does not denature protein samples
  • Stable and non-volatile in the electron beam
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
pH 8
Quantity 5 mL
Grade Cryo-EM grade
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.