Learn More
Vitronectin Antibody (019), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90648MFV450
Product Code. 18436339
  • € 674.00 / 0.10mL
Description
Vitronectin Monoclonal antibody specifically detects Vitronectin in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELISA, Immunohistochemistry (Paraffin)
ELISA, Immunohistochemistry-Paraffin
7448
IgG
019
50mM Sodium Borate
Store at 4C in the dark.