missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slow Skeletal Muscle Myosin Heavy Chain Antibody (M14), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50297AF350
This item is not returnable.
View return policy
Description
Slow Skeletal Muscle Myosin Heavy Chain Monoclonal specifically detects Slow Skeletal Muscle Myosin Heavy Chain in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Slow Skeletal Muscle Myosin Heavy Chain | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| purified human skeletal muscle myosin | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| M14 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| Cardiovascular Biology, Cellular Signaling, Developmental Biology, Stem Cells | |
| Specific for slow skeletal muscle myosin heavy chain (SM1). | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction