missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Vitronectin Monoclonal Antibody (347317)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA524083
This item is not returnable.
View return policy
Description
In direct ELISAs, no cross-reactivity with recombinant human Vitronectin is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
VTN is a member of the pexin family. This protein is found in serum and tissues and promotes cell adhesion and spreading, inhibits the membrane-damaging effect of the terminal cytolytic complement pathway, and binds to several serpin serine protease inhibitors. The protein is a secreted protein and exists in either a single chain form or a clipped, two chain form held together by a disulfide bond.
Specifications
| Vitronectin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P29788 | |
| VTN | |
| Mouse myeloma cell line NS0-derived recombinant mouse Vitronectin Asp20-Lys478. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Frozen) | |
| 347317 | |
| Unconjugated | |
| VTN | |
| Aa1018; AI256434; complement S-protein; epibolin; extracellular matrix protein; glycoprotein 66; serum spreading factor; serum-spreading factor; somatomedin B; Somatomedin-B; S-protein; V75; Vitronectin; vitronectin (serum spreading factor, somatomedin B, complement S-protein); Vitronectin V10 subunit; Vitronectin V65 subunit; VN; VNT; VTN | |
| Rat | |
| Protein A/G | |
| RUO | |
| 22370 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction