Learn More
Invitrogen™ Vitronectin Monoclonal Antibody (A18)
Mouse Monoclonal Antibody
Brand: Invitrogen™ CSI0041802
Description
CSI 004-18-02 is highly specific for vitronectin. There is no evidence for cross-reactivity with other connective tissue proteins (fibronectin, elastin, collagen, laminin). CSI 004-18-02 cross-reacts with rabbit and horse vitronectin, no reactivity with human or sheep. CSI 004-18-02 has been successfully used in ELISA and Immunohistochemistry techniques. CSI 004-18-02 is suitable in ELISA and immunostaining of frozen PLP-fixed sections of bovine tissues. The antibody inhibits integrin-mediated cell adhesion to bovine vitronectin. It can be used to probe vitronectin conformation. CSI 004-18-02 reacts weakly in ELISA with vitronectin coated directly onto the microtiter plate. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
VTN is a member of the pexin family. This protein is found in serum and tissues and promotes cell adhesion and spreading, inhibits the membrane-damaging effect of the terminal cytolytic complement pathway, and binds to several serpin serine protease inhibitors. The protein is a secreted protein and exists in either a single chain form or a clipped, two chain form held together by a disulfide bond.
Specifications
| Vitronectin | |
| Monoclonal | |
| 1.03 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| P22458 | |
| VTN | |
| Lysed bovine corneal endothelial cells and extracellular matrix. | |
| 400 μL | |
| Primary | |
| Rabbit, Bovine, Horse | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| A18 | |
| Unconjugated | |
| VTN | |
| Aa1018; AI256434; complement S-protein; epibolin; extracellular matrix protein; glycoprotein 66; serum spreading factor; serum-spreading factor; somatomedin B; Somatomedin-B; S-protein; V75; Vitronectin; vitronectin (serum spreading factor, somatomedin B, complement S-protein); Vitronectin V10 subunit; Vitronectin V65 subunit; VN; VNT; VTN | |
| Mouse | |
| Protein A | |
| RUO | |
| 100009128, 100059034, 507525 | |
| 4°C, store in dark | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.