missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VKORC1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97577H
This item is not returnable.
View return policy
Description
VKORC1 Polyclonal antibody specifically detects VKORC1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| VKORC1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| EC 1.1.4.1, EDTP308, FLJ00289, IMAGE3455200, MST576, phylloquinone epoxide reductase, vitamin K dependent clMST134, vitamin K epoxide reductase complex subunit 1, vitamin K epoxide reductase complex, subunit 1, Vitamin K1 2,3-epoxide reductase subunit 1, vitamin K1 epoxide reductase (warfarin-sensitive), VKCFD2, VKORMGC2694 | |
| Produced in rabbits immunized with E. coli-derived Human VKORC1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| HRP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 79001 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction