missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VLDLR Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99696-100ul
This item is not returnable.
View return policy
Description
VLDLR Polyclonal antibody specifically detects VLDLR in Human samples. It is validated for ELISA
Specifications
| VLDLR | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL receptor, VLDL-R, VLDLRCH | |
| Produced in rabbits immunized with purified, recombinant Human VLDLR (Accession#: NP_003374.3; Met 1-Ser 797) | |
| 100 μg | |
| Apoptosis, Cholesterol Metabolism, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 7436 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction