missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VLDLR Antibody (VLDLR/1337), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53351-100UG
This item is not returnable.
View return policy
Description
VLDLR Monoclonal specifically detects VLDLR in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| VLDL R | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
| CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL receptor, VLDL-R, VLDLRCH | |
| Mouse | |
| 152 kDa | |
| 100 μg | |
| Apoptosis, Cholesterol Metabolism, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 7436 | |
| Store at 4C. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| VLDLR/1337 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| VLDLR | |
| Recombinant human VLDLR fragment from c-terminal (exact sequence is proprietary) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction