Learn More
VMAT2 Antibody (899327), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB8327MFV610
Product Code. 18500709
  • € 674.00 / 0.10mL
Description
VMAT2 Monoclonal antibody specifically detects VMAT2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VMAT2
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
0.1 mL
Neuroscience
6571
Store at 4C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
899327
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready
MGC120477, monoamine neurotransmitter transporter, Monoamine transporter, solute carrier family 18 (vesicular monoamine), member 2, Solute carrier family 18 member 2, SVAT, SVMTMGC120478, synaptic vesicular amine transporter, VAT2MGC26538, vesicle monoamine transporter type 2, vesicle monoamine/H+ antiporter, Vesicular amine transporter 2, VMAT2
NS0 mouse myeloma cell line transfected with human VMAT2, Met1-Asp514, Accession # Q05940