Learn More
VNN2 Antibody (3F8F8H6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06441-100ul
This item is not returnable.
View return policy
Description
VNN2 Monoclonal antibody specifically detects VNN2 in Human samples. It is validated for Flow Cytometry, ELISA, Sandwich ELISA Capture
Specifications
| VNN2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Sandwich ELISA Capture | |
| 3F8F8H6 | |
| Flow Cytometry 1:25-1:100, ELISA 1:1000-1:2000, Sandwich ELISA Capture 1:250-1:2000 | |
| EC 3.5.1, EC 3.5.1.92, FOAP-4, Glycosylphosphatidyl inositol-anchored protein GPI-80, GPI-80, GPI-80 variant protein 1, GPI-80 variant protein 2, GPI-80 variant protein 3, GPI-80 variant protein 4, pantetheinase, Protein FOAP-4, vanin 2, vanin-2, Vannin 2, vascular non-inflammatory molecule 2 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human VNN2 (Accession#: NP_004656.2; Met 1-Ser 492). | |
| 100 μg | |
| Endocrinology, Signal Transduction, Stem Cells | |
| 8875 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |