missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VNN3 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81875C
This item is not returnable.
View return policy
Description
VNN3 Polyclonal antibody specifically detects VNN3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| VNN3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 55350 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| HSA238982, MGC171203, vanin 3 | |
| VNN3 antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human VNN3. The immunogen is located within the last 50 amino acids of VNN3. Amino Acid Squence: RHYEISRDGRLRSRSGAPL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction