missing translation for 'onlineSavingsMsg'
Learn More

Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 1 Mark and Amber Graduations

Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 1 Mark and Amber Graduations allow accurate measurements of a volume of solution.

Brand:  Poulten & Graf GmbH 1 2304504

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15251139

  • € 56.43 / Pack of 5


Estimated Shipment
18-09-2020
Add to basket

Description

Description

  • With 1 mark
  • Amber graduations
  • Conformity approved
  • With batch certificate
Specifications

Specifications

EN ISO 648
No
Amber
5
One-mark Bulb Pipette
Non-sterile
AS
Yes
Ring marks
0.05 mL
Soda Lime Glass
50 mL
Videos
Documents