missing translation for 'onlineSavingsMsg'
Learn More

Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 1 Mark and Amber Graduations

Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 1 Mark and Amber Graduations allow accurate measurements of a volume of solution.

Brand:  Poulten & Graf GmbH 1 2302904

Additional Details : Weight : 1.25000kg

 View more versions of this product

Product Code. 15932110

  • € 24.55 / Pack of 5
Estimated Shipment: 17-05-2024
to see stock.
Add to basket

Description

Description

  • With 1 mark
  • Amber graduations
  • Conformity approved
  • With batch certificate
Specifications

Specifications

EN ISO 648
Soda Lime Glass
Yes
Ring marks
0.01 mL
One-mark Bulb Pipette
AS
Non-sterile
Amber
5
3 mL
Reusable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers