Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Amber Graduations
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Amber Graduations allow accurate measurements of a volume of solution.
Brand:  Poulten & Graf GmbH 1 2323104
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10209291
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • With 2 marks
  • Amber graduations
  • Conformity approved
  • With batch certificate
Specifications
Specifications
EN ISO 648
Soda Lime Glass
Yes
Ring marks
0.015 mL
Two-mark Bulb Pipette
AS
Non-sterile
Amber
5
5 mL
Reusable