Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Amber Graduations
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Amber Graduations allow accurate measurements of a volume of solution.
Brand:  Poulten & Graf GmbH 1 2322904
Additional Details : Weight : 1.25000kg
 View more versions of this product
Product Code. 15942120
  • € 27.90 / Pack of 5
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
Description
Description
  • With 2 marks
  • Amber graduations
  • Conformity approved
  • With batch certificate
Specifications
Specifications
EN ISO 648
Soda Lime Glass
Yes
Ring marks
0.01 mL
Two-mark Bulb Pipette
AS
Non-sterile
Amber
5
3 mL
Reusable