Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Amber Graduations
Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with 2 Marks and Amber Graduations allow accurate measurements of a volume of solution.
Brand: Poulten & Graf GmbH 1 2324504
Additional Details : Weight : 1.25000kg
Description
- With 2 marks
- Amber graduations
- Conformity approved
- With batch certificate
Specifications
EN ISO 648 | |
Soda Lime Glass | |
Yes | |
Ring marks | |
0.05 mL | |
Two-mark Bulb Pipette |
AS | |
Non-sterile | |
Amber | |
5 | |
50 mL | |
Reusable |