Learn More
Poulten & Graf Ltd VOLAC FORTUNA™ Burette with PTFE Key and Single Certificate
Poulten & Graf Ltd VOLAC FORTUNA™ Burette with PTFE Key and Single Certificate accurately delivers known volumes of a liquid in titrations.
€ 79.60 - € 98.25
Specifications
Scale Color | Blue |
---|---|
Stopcock | Straight |
Stopcock Material | PTFE |
Type | Schellbach Burette |
Material | Borosilicate Glass |
Product Code | Brand | Capacity (Metric) | Tolerance | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Tolerance | Height (Metric) | Price | Quantity & Availability | |||
15509421
|
Poulten & Graf Ltd
RB9806/WAC/F |
25 mL | 0.03 mL | 820mm |
€ 79.60
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||
15519421
|
Poulten & Graf Ltd
RB9806/WAC/G |
25 mL | 0.05 mL | 620mm |
€ 98.25
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||
15529421
|
Poulten & Graf Ltd
RB9806/WAC/H |
50 mL | 0.05 mL | 820mL |
€ 79.60
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||
Description
- With straight stopcock
- With PTFE key in glass barrel
- Amber graduation
- Conformity approved
- With single certificate
Specifications
Blue | |
PTFE | |
Borosilicate Glass | |
Class AS | |
Amber |
Straight | |
Schellbach Burette | |
EN ISO 385 | |
No | |
25mL |